Learn More
Tocris Bioscience™ XL 388
Potent and selective mTOR inhibitor; antitumor
Manufacturer:  Tocris Bioscience™ 4893/50
 View more versions of this product
Catalog No. 48-935-0
Add to cart
This item is not returnable. View return policy
Description
Description
Potent and selective mTOR inhibitor (IC50 = 9.9 nM). Inhibits mTOR activity in an ATP-competitive manner. Exhibits >300-fold selectivity for mTOR over PI 3-K and a range of other kinases. Displays antitumor activity in athymic nude mice implanted with tumor xenografts.
Specifications
Specifications
XL 388
>98
50mg
XL 388
455.5