Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

XLF Mouse anti-Human, Clone: OTI3E5, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525312

Catalog No. PIMA525312



Description

Description

Double-strand breaks in DNA result from genotoxic stresses and are among the most damaging of DNA lesions. This gene encodes a DNA repair factor essential for the nonhomologous end-joining pathway, which preferentially mediates repair of double-stranded breaks. Mutations in this gene cause different kinds of severe combined immunodeficiency disorders.
Specifications

Specifications

XLF
Monoclonal
0.59 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9H9Q4
NHEJ1; XRCC4-like factor; nonhomologous end-joining factor 1; FLJ12610; XLF ; Cernunnos
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79840
Flow Cytometry, Western Blot
OTI3E5
Unconjugated
NHEJ1
Liquid
NHEJ1
Full length human recombit protein of NHEJ1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit