Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPA Mouse anti-Human, Clone: AT37C5, Abnova™

Mouse monoclonal antibody raised against full length recombinant human XPA.

Manufacturer:  Abnova Corporation MAB12476

Catalog No. 89-935-112

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

This gene encodes a zinc finger protein involved in DNA excision repair. The encoded protein is part of the NER (nucleotide excision repair) complext which is responsible for repair of UV radiation-induced photoproducts and DNA adducts induced by chemical carcinogens. Mutations in this gene are associated with xeroderma pigmentosum complementation group A. Alternatively spliced transcript variants have been found for this gene. [provided by RefSeq]

Specifications

Specifications

XPA
AT37C5
Mouse monoclonal antibody raised against full length recombinant human XPA.
In PBS, pH 7.4 (10% glycerol, 0.02% sodium azide)
XP1/XPAC
Mouse
IgG3, kappa
50 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
ELISA, Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
ELISA Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) Western Blot (1:500-1:2000) The optimal working dilution should be determined by the end user.
Liquid
XPA
Recombinant protein corresponding to full length of human XPA.
Protein A purification
RUO
Primary
7507
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit