Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPA Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen XPA
Applications Immunofluorescence, KnockDown, Western Blot
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093616
View Documents
Novus Biologicals
NBP303415100UL
100 μg
Each for $320.71
Add to cart
 
NB093617
View Documents
Novus Biologicals
NBP30341520UL
20 μg
Each for $138.37
Add to cart
 
Description

Description

XPA Polyclonal antibody specifically detects XPA in Human, Mouse samples. It is validated for Western Blot, Immunofluorescence, KnockDown
Specifications

Specifications

XPA
Unconjugated
Rabbit
DNA Repair, Nucleotide Excision Repair
PBS with 50% glycerol, pH7.3.
7507
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Immunofluorescence, KnockDown, Western Blot
Purified
RUO
Human, Mouse
DNA repair protein complementing XP-A cells, excision repair-controlling, Xeroderma pigmentosum group A-complementing protein, xeroderma pigmentosum, complementation group A, XPACXP1
Recombinant fusion protein containing a sequence corresponding to amino acids 1-273 of human XPA (NP_000371.1).
Polyclonal
Affinity purified
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit