Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPE/DDB2 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$149.00 - $329.00

Specifications

Antigen XPE/DDB2
Immunogen Synthetic peptides corresponding to DDB2 (damage-specific DNA binding protein 2, 48kDa) The peptide sequence was selected from the C terminal of DDB2. Peptide sequence IVVGRYPDPNFKSCTPYELRTIDVFDGNSGKMMCQLYDPESSGISSLNEF.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP16887920 View Documents Novus BiologicalsSupplier Diversity Partner
NBP16887920UL
20ul Each for $149.00
Add to cart
 
NBP168879 View Documents Novus BiologicalsSupplier Diversity Partner
NBP168879
100 ul Each for $329.00
Add to cart
 
Description

Description

XPE/DDB2 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

XPE/DDB2
Rabbit
Polyclonal
Unconjugated
1643
Affinity Purified
Western Blot
RUO
Synthetic peptides corresponding to DDB2 (damage-specific DNA binding protein 2, 48kDa) The peptide sequence was selected from the C terminal of DDB2. Peptide sequence IVVGRYPDPNFKSCTPYELRTIDVFDGNSGKMMCQLYDPESSGISSLNEF.
Primary
IgG
damage-specific DNA binding protein 2 (48kD), damage-specific DNA binding protein 2, 48kDa, Damage-specific DNA-binding protein 2, DDB p48 subunit, DDBb, DNA damage-binding protein 2, FLJ34321, UV-damaged DNA-binding protein 2, UV-DDB 2, UV-DDB2, xeroderma pigmentosum group E protein
PBS and 2% Sucrose with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit