Learn More
Mettler Toledo™ XPE-S Series Precision Balances Available on GSA/VA Contract for Federal Government customers only.
Provide outstanding weighing performance and support the highest requirements for safety, efficiency and ease of compliance
Manufacturer:  Mettler Toledo™ 30116495
 View more versions of this product
Catalog No. 01-911-573
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
  • Durable and easy to clean
  • High resolution technology with proFACT Advanced internal adjustment (on selected models)
  • SmartPan™ weighing pan with an integrated drip tray minimizes effects of air currents on weighing cell to deliver results up to two times faster
  • Repeatability values are improved up to two-fold
  • MonoblocHighSpeed weighing cell for guaranteed performance
  • StatusLight™ indicates intuitively the status of the balance by using colors
  • LevelGuide™ provides warning when the balance is not level
  • Small footprint, ergonomic space-saver design
  • Large color touchscreen with big numbers on display
  • SmartTrac™ dosing guide
  • Compact draft shield has all-round access for handling large items
  • ErgoStand protects the display from spills and makes it easier to read
  • SmartSens infra-red sensors built into the terminal for touch-less operation
  • TestManager embedded software
  • MinWeigh function
  • FACT and GWP history
  • LabX™ software provides flexible SOP user guidance on the balance touchscreen
  • Calibry ready
  • RFID exchange with titrators
  • RFID SampleTrack
  • Built in RS232 with seven options for the second slot
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
6.1kg; 1.2kg
RFID
N/A
5mg; 40mg
Solid metal housing; stainless-steel weighing pan
N/A
Precision Balance
UL Listed. CE compliant
60mg
XPE6002SDR/A
1.2sec
6.1kg; 1.2kg