Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPF Mouse anti-Human, Alexa Fluor™ 594, Clone: 4H4, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP253110AF594

Catalog No. NP23110A94


Add to cart

Description

Description

XPF Monoclonal antibody specifically detects XPF in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

XPF
4H4
Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Conjugated, Purified
ERCC4
XPF
Protein G purified
Cancer, DNA Repair, Nucleotide Excision Repair
Primary
2072.0
ELISA, Immunocytochemistry, Immunofluorescence, Western Blot
Alexa Fluor™ 594
50 mM sodium borate with 0.05% sodium azide
DNA excision repair protein ERCC-4, DNA repair protein complementing XP-F cells, EC 3.1, ERCC11, excision repair cross-complementing rodent repair deficiency, complementationgroup 4, xeroderma pigmentosum, complementation group F, XPFcomplementing defective, in Chinese hamster
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit