Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPF Mouse anti-Human, Biotin, Clone: OTI3H7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274882B

Catalog No. NBP274882B


Add to cart

Description

Description

XPF Monoclonal antibody specifically detects XPF in Human samples. It is validated for Western Blot.
Specifications

Specifications

XPF
OTI3H7
Western Blot
Affinity Purified, Conjugated
ERCC4
Full length recombinant protein of human XPF (NP_005227) produced in HEK293T cells.
Immunogen affinity purified
Cancer, DNA Repair, Nucleotide Excision Repair
Primary
2072.0
Western Blot
Biotin
PBS with 0.05% Sodium Azide
DNA excision repair protein ERCC-4, DNA repair protein complementing XP-F cells, EC 3.1, ERCC11, excision repair cross-complementing rodent repair deficiency, complementationgroup 4, xeroderma pigmentosum, complementation group F, XPFcomplementing defective, in Chinese hamster
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit