Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

XPF Mouse anti-Human, Clone: OTI1B2, liquid, TrueMAB™ Non-distribution product as customer accommodation.

Mouse Monoclonal Antibody

Manufacturer:  OriGeneSupplier Diversity Partner TA503262

Catalog No. 50-167-4201



Description

Description

The structure specific ERCC 1 / XPF endonuclease complex is implicated in the repair of two distinct types of lesions in DNA: NER for UV induced lesions and bulky chemical adducts; and recombination repair of the very genotoxic interstrand cross links. NER mechanism involves dual incisions on both sides of the damage catalyzed by two nucleases. In mammalian cells, XPG cleaves 3 ft. of the DNA lesion while the ERCC1 / XPF complex makes the 5 ft. incision. ERCC1 / XPF probably plays a role in processing partially processed recombination intermediates.
Specifications

Specifications

XPF
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q92889
ERCC11, FANCQ, RAD1, XFEPS, XPF
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
2072
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot
OTI1B2
Unconjugated
ERCC4
Liquid
ERCC4
Full length human recombit protein of human ERCC4 produced in HEK293T cell.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit