Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPF Mouse anti-Rat, FITC, Clone: 3F2/3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP127881F

Catalog No. NBP127881F


Add to cart

Description

Description

XPF Monoclonal antibody specifically detects Antigen in Rat samples. It is validated for Enzyme Linked Immunosorbent Assay, Western Blotting.
Specifications

Specifications

XPF
XPF 3F2/3
ELISA, Immunocytochemistry/Immunofluorescence
Purified
ERCC4
A recombinant His-tagged fragment of human XPF protein produced in E. coli
Protein A purified
RUO
Store at 4C in the dark.
Monoclonal
Human
ELISA, Immunocytochemistry, Immunofluorescence
FITC
PBS with 0.05% Sodium Azide
DNA excision repair protein ERCC-4, DNA repair protein complementing XP-F cells, EC 3.1, ERCC11, excision repair cross-complementing rodent repair deficiency, complementationgroup 4, xeroderma pigmentosum, complementation group F, XPFcomplementing defective, in Chinese hamster
Mouse
IgG1
0.2 ml
Cancer, DNA Repair, Nucleotide Excision Repair
Primary
2072
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit