Learn More
Eppendorf™ Xplorer™ 12-channel plus epT.I.P.S. Bundle
Eppendorf Xplorer 12-channel pipette is designed to aspirate and dispense liquid volumes from 0.5µL to 10 mL while eliminating many of potential error risks associated with manual pipetting.
Manufacturer:  Eppendorf™ 2231000328
 View more versions of this product
Catalog No. 05-412-721
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Intuitive operating concept for quick and easy work
  • Multifunctional rocker
  • Selection dial
  • Innovative ejector
  • Ergonomic display angle
  • Convenient handle
  • Powerful rechargeable battery
  • Flexible charging concept
  • Spring-loaded tip cone (except 5 mL and 10 mL)
  • Adjustment to external conditions (geographic height, liquids with high density, or similar factors)
Specifications
Specifications
50 to 1200 μL
epT.I.P.S
Green
Rechargeable
Digital
12