Learn More
Eppendorf™ Xplorer™ 8-channel plus epT.I.P.S. Bundle
Eppendorf Xplorer 8-channel pipette is designed to aspirate and dispense liquid volumes from 0.5μL to 10 mL while eliminating many of potential error risks associated with manual pipetting.
Specifications
Number of Channels 8
Battery Type Rechargeable
Compatible Tips epT.I.P.S
View More Specs
Products 1
Catalog Number Mfr. No. Volume (Metric) Color Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Color Price Quantity & Availability  
05-412-720
View Documents
Eppendorf™
2231000327
50 to 1200 μL Green This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Intuitive operating concept for quick and easy work
  • Multifunctional rocker
  • Selection dial
  • Innovative ejector
  • Ergonomic display angle
  • Convenient handle
  • Powerful rechargeable battery
  • Flexible charging concept
  • Spring-loaded tip cone (except 5 mL and 10 mL)
  • Adjustment to external conditions (geographic height, liquids with high density, or similar factors)
Specifications
Specifications
8
Rechargeable
epT.I.P.S