Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XPNPEP1 Mouse anti-Human, Clone: OTI1E7, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA525169

Catalog No. PIMA525169


Add to cart

Description

Description

This gene encodes the cytosolic form of a metalloaminopeptidase that catalyzes the cleavage of the N-terminal amino acid adjacent to a proline residue. The gene product may play a role in degradation and maturation of tachykinins, neuropeptides, and peptide hormones. Alternative splicing results in multiple transcript variants.
Specifications

Specifications

XPNPEP1
Monoclonal
0.58 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NQW7
XPNPEP1, X-prolyl aminopeptidase (aminopeptidase P) 1, soluble, SAMP, XPNPEP, XPNPEPL, XPNPEPL1, OTTHUMP00000020457, OTTHUMP00000058856, X-prolyl aminopeptidase (aminopeptidase P) 1, soluble (SAMP, XPNPEP, XPNPEPL), X-prolyl aminopeptidase (aminopeptidase P)-like
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7511
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
OTI1E7
Unconjugated
XPNPEP1
Liquid
XPNPEP1
Full length human recombit protein of XPNPEP1 produced in HEK293T cell
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit