Learn More
Mettler Toledo™ XPR Microbalances Available on GSA/VA Contract for Federal Government customers only.
Ensure traceable results with Active Temperature Control system and Tolerance Profiles. The Mettler Toledo XPR Microbalances and Ultra-Microbalances deliver unmatched accuracy in the micro-weighing range, compact space-saving design with features for safe operation as well as efficient process security. 
Manufacturer:  Mettler Toledo™ 30279285
 View more versions of this product
Catalog No. 01-914-322
Due to product restrictions, please to purchase or view availability for this product.
Description
Description
Laboratory projects involving expensive, rare or toxic substances demand that only the smallest possible sample quantities are used. Not only do you have the challenge to prevent waste by weighing out just a few milligrams, but you also need to avoid out-of-specification results and costly reworking. Mettler Toledo XPR Microbalances provide the following features:
  • Deliver a unique level of precision with exceptionally low minimum weights
  • Two-terminal concept simplifies operation: The main terminal can be placed wherever it is most convenient and the SmartView terminal sits above the weighing chamber – right where you need it - for all the basic functions you want as you carry out your weighing tasks
  • New draft shield provides easy access to the weighing pan; all parts are easily removed for cleaning
  • Cross-contamination risk is reduced thanks to SmartSens infrared sensors which enable you to operate the draft shield with the wave of a hand
  • Active Temperature Control system (ATC™)
  • Magnetic Force Restoration weighing sensor with high resolution
  • MinWeigh warning function
  • Capacitive colored touchscreen with gesture control, glove compatible
  • Method library saves time and provides higher process security
  • Built-in notepad automatically records all task parameters and results, eliminating manual transcription
  • Efficient operation via SmartView terminal
  • Storage of SOPs in method library (including sample series and tolerances)
  • Multiple ports for easy connectivity and data export: 4 x USB, 1 x LAN
  • Compact design with small footprint
  • Optimized draft shield for easy cleaning
  • Overload protection
Quality Assurance Features:
  • GWP Approved quality assurance function actively monitors the balance to confirm it is safe to weigh: tests, adjustments, levelling and settings must all be in order
  • Tolerance Profiles
  • Password protection
  • User management
  • Change history function
  • Adjustment and routine test history
  • Balance ready StatusLight™
  • LevelControl: Level warning with graphical levelling guide
  • Temperature and time programmable proFACT
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
0.003mg
Ultra Micro Balance
0.06 mg
27mm
0.0005 mg
< 8 sec.