Learn More
XRCC1 (X-Ray Repair Cross Complementing Protein 1) Ab-1 Mouse Monoclonal Antibody, Epredia™
Provide accurate, reproducible results with the Epredia™ XRCC1 (X-Ray Repair Cross Complementing Protein 1) Ab-1, Mouse Monoclonal Antibody.
$227.00 - $1189.00
Specifications
Antigen | XRCC1 (X-Ray Repair Cross Complementing Protein1) Ab-1 |
---|---|
Clone | 33-2-5 |
Applications | Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Monoclonal |
Host Species | Mouse |
Catalog Number | Mfr. No. | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Form | Conjugate | Quantity | Price | Quantity & Availability | |||
MS434P0
|
Epredia
MS434P0 |
Concentrated | Unconjugated | 100 μL |
Each for $227.00
|
|
|||
MS434P1ABX
|
Epredia
MS434P1ABX |
Concentrated | Unconjugated | 100 μL |
Each for $787.00
|
|
|||
MS434PABX
|
Epredia
MS434PABX |
Concentrated | Unconjugated | 200 μL |
Each for $1,189.00
|
|
|||
MS434P1
|
Epredia
MS434P1 |
Concentrated | Unconjugated | 500 μL |
Each for $949.00
|
|
|||
MS434R7
|
Epredia
MS434R7 |
Prediluted | Unconjugated | 7 mL |
Each for $676.00
|
|
|||
MS434B0
|
Epredia
MS434B0 |
Concentrated | Biotin | 100 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS434B1
|
Epredia
MS434B1 |
Concentrated | Biotin | 500 μL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS434B
|
Epredia
MS434B |
Concentrated | Biotin | 1 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS434P
|
Epredia
MS434P |
Concentrated | Unconjugated | 1 mL | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||
MS434R1
|
Epredia
MS434R1 |
Prediluted | Unconjugated | 1 mL |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | |||
Description
X-Ray Repair Cross Complementing (XRCC1) plays a role in excision repair of DNA after ionizing irradiation. XRCC1 binds to DNA ligase III through the C-terminal 96 amino acids and to DNA polymerase β through the N-terminal half. In testis XRCC1 is expressed at high levels. Cells with a mutation of this gene exhibit decreased single strand break repair and reduced recombination repair. They show increased double strand breaks, and sister chromatid exchange is increased up to 10-fold. The XRCC1 might serve as a scaffold protein during base excision-repair.Host Species: Mouse
Clone: 33-2-5
Isotype: IgG2b
Species Reactivity: Human and Rat. Others not known.
Epitope: Not determined
Immunogen: Recombinant human XRCC1 protein
Molecular Weight: 85kDa
Positive Control: Ls174T, HT29 or HeLa cells; normal testis
Cellular Localization: Nuclear
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
- Immunoprecipitation (Native verified)
Note:
USA: RUO; Int'l: RUO
Specifications
XRCC1 (X-Ray Repair Cross Complementing Protein1) Ab-1 | |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Mouse | |
DNA Repair | |
Recombinant human XRCC1 protein | |
Primary |
33-2-5 | |
Monoclonal | |
RUO | |
Human, Rat | |
IgG2b |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.