Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ XRCC3 Recombinant Protein
Specifications
Accession Number | AAH01036.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Gene ID (Entrez) | 7517 |
Molecular Weight (g/mol) | 63.69 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-011-088
|
Abnova™
H00007517P0110 |
10 ug | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
89-011-089
|
Abnova™
H00007517P0125 |
25 ug | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | |||||
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH01036.1 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
63.69 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
XRCC3 | |
Wheat Germ (in vitro) | |
GST |
Antibody Production, Protein Array, ELISA, Western Blot | |
7517 | |
XRCC3 (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MDLDLLDLNPRIIAAIKKAKLKSVKEVLHFSGPDLKRLTNLSSPEVWHLLRTASLHLRGSSILTALQLHQQKERFPTQHQRLSLGCPVLDALLRGGLPLDGITELAGRSSAGKTQLALQLCLAVQFPRQHGGLEAGAVYICTEDAFPHKRLQQLMAQQPRLRTDVPGELLQKLRFGSQIFIEHVADVDTLLECVNKKVPVLLSRGMARLVVIDSVAAPFRCEFDSQASAPRARHLQSLGATLRELSSAFQSPVLCINQVTEAMEEQGAAHGPLGFWDERVSPALGITWANQLLVRLLADRLREEEAALGCPARTLRVLSAPHLPPSSCSYTISAEGVRGTPGTQSH | |
XRCC3 | |
Human | |
Recombinant | |
Solution |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title