Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XRCC4 Mouse anti-Human, DyLight™ 350, Clone: OTI4H9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274896UV

Catalog No. NBP274896UV


Add to cart

Description

Description

XRCC4 Monoclonal antibody specifically detects XRCC4 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

XRCC4
OTI4H9
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
XRCC4
Full-length protein expressed in 293T cell transfected with human XRCC4 expression vector.
Immunogen affinity purified
Cancer, DNA Double Strand Break Repair, DNA Repair, Homologous Recombination, Non homologous end joining, Tumor Suppressors
Primary
7518.0
Immunohistochemistry, Western Blot
DyLight 350
50mM Sodium Borate with 0.05% Sodium Azide
complementing defective, repair in Chinese hamster, X-ray repair complementing defective repair in Chinese hamster cells 4, X-ray repair cross-complementing protein 4
Mouse
IgG1
0.1 mL
Store at 4°C in the dark.
Monoclonal
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit