Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XRCC5 Mouse anti-Human, Clone: 9403, Abnova™

Mouse monoclonal antibody raised against partial recombinant human XRCC5.

Manufacturer:  Abnova Corporation MAB12933

Catalog No. 89-935-115


Add to cart

Description

Description

The protein encoded by this gene is the 80-kilodalton subunit of the Ku heterodimer protein which is also known as ATP-dependant DNA helicase II or DNA repair protein XRCC5. Ku is the DNA-binding component of the DNA-dependent protein kinase, and it functions together with the DNA ligase IV-XRCC4 complex in the repair of DNA double-strand break by non-homologous end joining and the completion of V(D)J recombination events. This gene functionally complements Chinese hamster xrs-6, a mutant defective in DNA double-strand break repair and in ability to undergo V(D)J recombination. A rare microsatellite polymorphism in this gene is associated with cancer in patients of varying radiosensitivity. [provided by RefSeq]

Specifications

Specifications

XRCC5
9403
Mouse monoclonal antibody raised against partial recombinant human XRCC5.
In PBS.
FLJ39089/KARP-1/KARP1/KU80/KUB2/Ku86/NFIV
Mouse
IgG
50 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Monoclonal
Human
Immunohistochemistry (PFA fixed), Western Blot
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (5 ug/mL) Western Blot The optimal working dilution should be determined by the end user.
Liquid
XRCC5
Recombinant protein corresponding to N-terminus of human XRCC5.
Protein G purification
RUO
Primary
7520
SDS
Documents
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit