Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
XRCC6BP1 Mouse anti-Human, FITC, Clone: OTI5F3, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP274897F
Description
XRCC6BP1 Monoclonal antibody specifically detects XRCC6BP1 in Human samples. It is validated for Immunohistochemistry.Specifications
XRCC6BP1 | |
OTI5F3 | |
Immunohistochemistry | |
Affinity Purified, Conjugated | |
ATP23 | |
Full length recombinant protein of human XRCC6BP1 (NP_150592) produced in E.coli. | |
Immunogen affinity purified | |
Protein Kinase | |
Primary | |
91419.0 |
Immunohistochemistry | |
FITC | |
PBS with 0.05% Sodium Azide | |
EC 3.4.24, EC 3.4.24.-, Ku70-binding protein 3, KUB3Ku70 binding protein 3, MGC134817, MGC134818, mitochondrial inner membrane protease ATP23 homolog, XRCC6 binding protein 1, XRCC6-binding protein 1 | |
Mouse | |
IgG1 | |
0.1 mL | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title