Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XRCC6BP1 Mouse anti-Human, Clone: OTI5F3, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274897

Catalog No. NBP274897


Add to cart

Description

Description

XRCC6BP1 Monoclonal antibody specifically detects XRCC6BP1 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

XRCC6BP1
OTI5F3
Unconjugated
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
EC 3.4.24, EC 3.4.24.-, Ku70-binding protein 3, KUB3Ku70 binding protein 3, MGC134817, MGC134818, mitochondrial inner membrane protease ATP23 homolog, XRCC6 binding protein 1, XRCC6-binding protein 1
Mouse
IgG1
Immunogen affinity purified
Protein Kinase
Primary
91419
Human
Immunohistochemistry, Western Blot
LYOPH
Western Blot 1:500, Immunohistochemistry 1:150
Affinity Purified, Lyophilized
ATP23
Full length human recombinant protein of human XRCC6BP1 (NP_150592) produced in E.coli.
27.9 kDa
100 μg
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit