Learn More
XRCC6BP1 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 160761AP
Catalog No. 50-173-0694
Add to cart
Description
Description
The protein encoded by this gene is amplified in glioblastomas and interacts with the DNA binding subunit of DNA-dependent protein kinase. This kinase is involved in double-strand break repair (DSB), and higher expression of the encoded protein increases the efficiency of DSB. In addition, comparison to orthologous proteins strongly suggests that this protein is a metalloprotease important in the biosynthesis of mitochondrial ATPase. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
XRCC6BP1
0.33 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Q9Y6H3
ATP23, Ku70 binding protein 3, KUB3, XRCC6 binding protein 1, XRCC6BP1
Rabbit
IgG
150 μL
-20°C
Primary
91419
Western Blot
Unconjugated
ATP23
Liquid
ATP23
XRCC6BP1 Fusion Protein Ag9042
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human