Learn More
Waters Corp XSelect CSH C18 XP Column, 130Å, 2.5 µm, 3 mm X 30 mm, 3/pk
Supplier:  Waters Corp 176002598
Encompass_Preferred
XSelect CSH C18 columns are general purpose C18 columns that provide alternative selectivity compared to other C18 sorbents. Charged Surface Hybrid (CSH) technology enables superior peak shape and increased loading capacity for basic compounds. Packed in ultra-low dispersion hardware, 2.5 µm eXtended Performance [XP] columns enable exceptional separation performance, robustness and throughput for HPLC assays while enabling a seamless transition path toward UPLC adoption.
Catalog No. 50-179-257
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.