Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

METTLER TOLEDO INC XSR Precision Balance

METTLER TOLEDO INC XSR Precision Balances offer reliable performance and ergonomic operation even under the harshest of weighing conditions

Manufacturer:  METTLER TOLEDO INC 30317540

 View more versions of this product

Catalog No. 01-919-087


Due to product restrictions, please to purchase or view availability for this product.


Description

Description

Features:

  • Easy and comfortable to use, protecting users' health during your daily weighing tasks
  • Stability provided by the Monobloc™ weighing cell produces results quickly even under challenging production environments
  • NTEP Approved
  • Large pan balances
  • 4.3 inch color capacitive touchscreen terminal, glove compatible
  • Storage of weighing methods
  • Integrated results notepad
  • Multiple interfaces for easy connectivity and data export: 4 x USB, 1 LAN
  • Easy connection of peripheral devices: printers, PC, barcode readers, footswitches and sensors
  • Export of process and results data to XML or Excel
  • Customizable tolerance profiles, with pre-defined quality and weighing settings
  • MinWeigh warning function
  • Balance ready StatusLight™
  • LevelControl: level warning with graphical leveling guide
  • Temperature and time programmable automatic adjustment (proFACT)
  • Password protected configuration
  • Integrated overload protection
  • Chemically resistant materials
  • Designed for easy cleaning

Warranty and Services

Yes

Specifications

Specifications

4 x USB, LAN
0.1 g
Die-cast Zinc, Die-cast Aluminum
100-240 V AC, 50/60 Hz
4.3-inch Color Touchscreen
NTEP Approved, Legal for Trade
XSR10001L/A
1.5 s
10100 g
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

*For use with Legal for Trade / Approved Model Applications*