Learn More
Corning™ XT Freezing Core for CoolBox™ XT Workstation
Allows for continuous uninterupted cooling. Corning™ XT Freezing Core for CoolBox™ XT Workstation provides cooling source in CoolBox XT Workstation when thermo-conductive CoolRack or CoolSink tube and plate module is placed on top.
Manufacturer:  Corning™ 432082
Catalog No. 07-210-082
Add to cart
Description
Description
XT Freezing Core for CoolBox XT, for below 0°C, a second one allows for continuous, uninterupted cooling.
Specifications
Specifications
XT Freezing Core
1/Pk.
Maintaining sample temperature
16.2 x 12.1 x 6.1 cm (6.3 x 4.7 x 2.1 in.)
Blue
1/Cs.
CoolBox XT Workstation