Corning™ XT Starter Ice-free Coolers

Bench top cooling systems that provide sample cooling or freezing without ice, electricity, or batteries.

Manufacturer: Corning™ 432016

 View more versions of this product

Catalog No. 07-210-016

  Check Availability
Add to Cart

    Specifications

    Quantity 1
    Product Type Ice-free cooler
    Holds One 96-well PCR plate, strip-wells or 200μL tubes
    Color Purple
    Includes One XT holder base, one XT Cooling Core and a CoolRack PCR module
    For Use With (Application) Sample cooling or freezing without ice, electricity, or batteries
    Dimensions (L x W x H) Exterior 16.2 x 12.0 x 8.2cm

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit