Learn More
Biotix™ xTIP™ CleanPak™ Low Retention Pipette Tips for Rainin™ LTS™ Style Pipettors
Designed with hermetically sealed bag that makes contamination a thing of the past. Biotix™ xTIP™ CleanPak™ Low Retention Pipette Tips for Rainin™ LTS™ style pipettors reduce time to rack product, cost, and environmental footprint.
Manufacturer:  Biotix™ R02509TS
 View more versions of this product
Catalog No. 12-111-043
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description
Description
  • Designed for Rainin LTS-Style Pipets with Biotix technology
  • FlexFit, X-Resin&trade, and Blade technologies enhance every aspect of these tips
  • Ensure clean, accurate delivery
  • 960 tips on lab bench using the same surface area as 1 rack
  • Sterile and non-Sterile re-sealable bag available
  • Clear bag and color coded snap cards for quick identification
  • Fully recyclable
  • Contains no contaminating cardboard or paper
  • Inserts are easy to transfer
Certifications
ISO 9001 , FDA Class I
Specifications
Specifications
250 μL
Non-filtered
Sterile
Refill