Learn More
Biotix™ xTIP™ Eco-Friendly Empty Racks with Removable Lid
Manufactured using 30-60% less plastic than competitor racks. Biotix™ xTIP™ Eco-Friendly Empty Racks with Removable Lid are compatible with reload options including Biotix TipEject™ and CleanPak™.
Specifications
No. per Case 4 Pks./Cs.
No. per Pack 10 Racks/Pk.
Quantity 960 Tips/Pk., 4 Pks./Cs.
View More Specs
Products 2
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
12-111-051 View Documents Biotix™
R00209ER
N/A Vol.: 10-250µL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
12-111-052 View Documents Biotix™
R10009ER
N/A Vol.: 1000µL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Features pyramidal wells on base for use as reservoir
Certifications
ISO 9001 , FDA Class I
xTIP, EmptyRack, Reload, CleanPak, TipEject, Biotix, Liftoff, Eco-friendly, automation, SBS, reservoir
Specifications
Specifications
4 Pks./Cs.
10 Racks/Pk.
960 Tips/Pk., 4 Pks./Cs.