Learn More
Biotix™ xTIP™ Low-Retention Filter Pipette Tips
Manufactured with X-Resin™ low retention resin tightening CV values. Biotix™ xTIP™ Low-Retention Filter Pipette Tips are designed with Flexfit™ to lower insertion and ejection forces to make secure seal.
Specifications
Graduated Yes
Sterility Sterile
Material PP (Polypropylene)
View More Specs
Products 3
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
12-111-033
View Documents
Biotix™
R00209FC
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
12-111-034
View Documents
Biotix™
R02009FC
200 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
12-111-035
View Documents
Biotix™
R10009FC
1000 μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
  • Designed for Rainin LTS-Style Pipets with Biotix technology
  • FlexFit, X-Resin, and Blade technologies enhance every aspect of these tips
  • Ensure clean, accurate delivery every time
  • FlexFit technology features flexible proximal end to ensure secure seal with 66-80% less insertion force than competitor tips
  • X-Resin is proprietary low retention resin proven to drastically reduce sample retention resulting in tighter CV values
  • Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip. Blade eliminates need for tip touch off
  • Pre-Sterile products undergo E-Beam radiation
Certifications
ISO 9001 , FDA Class I
Specifications
Specifications
Yes
Sterile
PP (Polypropylene)