Learn More
Biotix™ xTIP™ Low Retention, Racked Pipette Tips for Rainin™ LTS™ Style Pipettors
Manufactured with X-Resin™ low retention resin tightening CV values. Biotix™ xTIP™ Low Retention, Racked Pipette Tips for Rainin™ LTS™ style pipettors are designed with Flexfit™ to lower insertion and ejection forces to make secure seal.
Manufacturer:  Biotix™ R00209NC
 View more versions of this product
Catalog No. 12-111-039
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Designed for Rainin LTS-Style Pipets with Biotix technology
  • FlexFit, X-Resin, and Blade technologies enhance every aspect of these tips
  • Ensure clean, accurate delivery every time
  • FlexFit technology features flexible proximal end to ensure secure seal with 66-80% less insertion force than competitor tips
  • X-Resin is proprietary low retention resin proven to drastically reduce sample retention resulting in tighter CV values
  • Blade technology lowers chance of hanging droplet formation by optimizing surface area at distal end of tip. Blade eliminates need for tip touch off
  • Pre-Sterile products undergo E-Beam radiation
Certifications
ISO 9001 , FDA Class I
Specifications
Specifications
20 μL
Non-filtered
Rack