Learn More
Biotix™ xTIP™ TipEject™ Low Retention Pipette Tips for Rainin™ LTS™ Style Pipettors
For use in labs that need hands free option for reloads. Biotix™ xTIP™ TipEject™ Low Retention Pipette Tips for Rainin™ LTS™ style pipettors allow racking tips in seconds, and keeps tips shielded from environment.
Specifications
Sterility Sterile
Filtered Non-Filtered
View More Specs
Products 3
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
12-111-048 View Documents Biotix™
R00209RC
20μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
12-111-049 View Documents Biotix™
R02509RC
250μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
12-111-050 View Documents Biotix™
R10009RC
1000μL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Designed for Rainin LTS-Style Pipets with Biotix technology
  • FlexFit, X-Resin™ and Blade technologies enhance every aspect of these tips
  • Ensure clean, accurate delivery
  • 960 tips on lab bench using the same surface area as 1 rack
  • Sterile and non-Sterile re-sealable bag available
  • Clear bag and color coded snap cards for quick identification
  • Fully recyclable
  • Contains no contaminating cardboard or paper
  • Inserts are easy to transfer
Certifications
ISO 9001 , FDA Class I
xTIP, RaininLTS, TipEject, Reload, 10 cards/pack, Low-Retention, flexfit, blade, x-resin
Specifications
Specifications
Sterile
Non-Filtered