Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XTP4 Mouse anti-Human, Clone: OTI4D4, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA526354

Catalog No. PIMA526354


Add to cart

Description

Description

During the past decade, the role of the ERBB2(neu/HER2) oncogene as an important predictor of patient outcome and response to various therapies in breast cancer has been clearly established. The C35 (C17orf37) is a novel tumor biomarker abundantly expressed in breast cancer. Identification of shared tumor-specific targets is useful in developing broadly applicable therapies. The C35 gene is located on chromosome 17q12, 505nucleotides from the 3 ft. end of the ERBB2oncogene, the antigenic target for trastuzumab(Herceptin) therapy. The chromosomal arrangement of the genes encoding C35 andERBB2 is tail to tail. The ORF 37 open reading frame encodes a 12-kDa protein of unknown function.
Specifications

Specifications

XTP4
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q9BRT3
C35, MGC14832, ORB3, RDX12, XTP4
Mouse
IgG2a
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84299
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
OTI4D4
Unconjugated
MIEN1
Liquid
MIEN1
Full length human recombit protein of C17orf37 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit