Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Kimberly-Clark™ Kimtech™ PFE-Xtra Latex Ambidextrous Gloves

Robust Kimtech™ PFE-Xtra Latex Ambidextrous Gloves are suitable for challenging applications and environments that have a high risk of chemical splash. The gloves are ambidextrous, non-sterile and made from skin-friendly natural rubber latex.

Manufacturer:  Kimberly-Clark™ 50502

 View more versions of this product

Catalog No. 19-048-565C



Add to cart

Description

Description

  • PFE-Xtra™ latex material provides high levels of contamination protection, including enhanced chemical splash.
  • Gloves are non-sterile, ambidextrous, disposable and white in color.
  • Textured palm and fingertips enhance grip and tactile sensitivity for safer and more efficient processes.
  • Beaded cuffs add strength to the gloves, reducing the risk of tearing and increasing their durability, while also reducing roll down for easier donning and doffing.
  • Made from natural rubber latex for a low dermatitis risk and containing no powder, reducing the risks of skin irritation for the wearer.
  • Assured Compliance:
    • PPE Cat III according to Regulation (EU) 2016/425
    • EN ISO 374-1:2016 Type C (K) Chemical Splash protection
    • EN 374-4:2003 Resistance to degradation by chemicals
    • EN ISO 374-5:2016 Micro Organism and VIRUS Protection
Specifications

Specifications

Safety
Yes
Textured
Ambidextrous
No
Chemical resistance
50Pack
Latex gloves
Natural
EN ISO 374-1:2016; EN 374-4:2003; EN ISO 374-5:2016
PPE Category II
Textured
Beaded/Rolled
Yes
Latex
Medium
0.21 (fingers, palm), 0.19 to 0.14 mm (cuff)
Powder-free
304.8 mm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit