Learn More
R&D Systems™ XTT Activator
R&D Systems™ XTT Activator is for use with TACS XTT Cell Proliferation Assay.
Manufacturer:  R&D Systems™ 489102502
Catalog No. 48-910-2502
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
This item is not returnable. View return policy
Description
Description
  • Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
  • Do not use past expiration date.
  • Contains PMS (phenazine methosulfate, CAS # 299- 11-6).
  • PMS is toxic and may be carcinogenic and/or mutagenic.
  • PMS is an irritant. In case of contact, immediately flush eyes or skin with copious amounts of water.
  • If swallowed, wash out mouth with water provided person is conscious.
  • The stock is 50X in dH2O. A volume of 100µL is sufficient for each 5mL of XTT reagent (i.e. one microplate).
  • Like the XTT reagent, the XTT activator will precipitate during storage.
  • Heat to 37°C for 2 to 5 minutes until the reagent is fully dissolved.
Specifications
Specifications
2.5mL
Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.