Learn More
Medchemexpress LLCSupplier Diversity Partner XY1 10MG DFS Item
Manufacturer:  Medchemexpress LLCSupplier Diversity Partner HY1971410
XY1 10MGCAS No. : 1624117-53-8XY1 is a very close analogue of SGC707 (a potent, selective, and non-competitive inhibitor of PRMT3 with IC50 of 31 nM), but XY1 is completely inactive.
Catalog No. NC1479205