Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylazine Hydrochloride, USP, 98-102%, Spectrum™

Click to view available options
Quantity:
1 g
5 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
23076-35-9
C12H17ClN2S
256.79
QYEFBJRXKKSABU-UHFFFAOYSA-N
hydrogen N-(2,6-dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine chloride
[H+].[Cl-].CC1=CC=CC(C)=C1NC1=NCCCS1
23076-35-9
C12H17ClN2S
256.79
QYEFBJRXKKSABU-UHFFFAOYSA-N
hydrogen N-(2,6-dimethylphenyl)-5,6-dihydro-4H-1,3-thiazin-2-amine chloride
[H+].[Cl-].CC1=CC=CC(C)=C1NC1=NCCCS1

X1067, 23076-35-9, C12H16N2S•HCl

Spectrum™ Xylazine Hydrochloride, USP is a common drug that is used for the sedation, anesthesia, muscle relaxation, and analgesia in many animals such as horses, cattle, and other non-human mammals. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Packaging Amber Glass Bottle
Ignition Residue 0.001
Quantity 25 g
Melting Range 164-168°C
Loss on Drying 0.01
CAS Max % 1
Percent Purity 98 to 102%
Grade USP