Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylene Cyanol FF, OmniPur™, Calbiochem™,

Click to view available options
Quantity:
25g
Packaging:
Plastic Bottle
2650-17-1
2650-17-1
Physical Form Powder
Grade Molecular Biology
Packaging Plastic Bottle
Color Blue-Green
Quantity 25g