Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenes, 96%, pure, mixed isomers with ethylbenzene, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
25L
Packaging:
Glass Bottle
Metal drum
1330-20-7,100-41-4
C8H10
MFCD00077264
Dimethylbenzene
1330-20-7,100-41-4
C8H10
MFCD00077264
Dimethylbenzene
Additional Information Vapor Pressure: 8mbar at 20°C
Assay Percent Range 96%
Linear Formula C6H4(CH3)2
Beilstein 05, 360
Merck Index 15, 10278
Formula Weight 106.17
Physical Form Liquid
Percent Purity 96%
Grade Pure
Density 0.8650g/mL
Infrared Spectrum Authentic
Packaging Metal drum
Refractive Index 1.4940 to 1.4980 (20°C, 589nm)
Solubility Solubility in water: 0.1g/L water (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf, methylene chloride, acetone and ether
Specific Gravity 0.865
Flash Point 23°C
Viscosity 0.6 mPa.s (20°C)
Boiling Point 136.0°C to 140.0°C
Color Undesignated
Melting Point -34.0°C
Quantity 25L
Chemical Name or Material Xylenes, Mixed Isomers with Ethylbenzene
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 215-535-7

RTECSNumber : ZE2100000

TSCA : TSCA