Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenes, 98+%, for analysis, mixture of isomers , ACROS Organics™

Click to view available options
Quantity:
1L
5L
1330-20-7,100-41-4
C8H10
Dimethylbenzene
1330-20-7,100-41-4
C8H10
Dimethylbenzene
Assay Percent Range 98+%
Linear Formula C6H4(CH3)2
Solubility Information Solubility in water: 0.1g/L water (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf, methylene chloride, acetone and ether
Formula Weight 106.17
Percent Purity 98+%
Grade Analysis
Density 0.8650g/mL
Solubility Solubility in water: 0.1g/L water (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf, methylene chloride, acetone and ether
Specific Gravity 0.865
Flash Point 23°C
Viscosity 0.6 mPa.s (20°C)
Boiling Point 136.0°C to 140.0°C
Melting Point -34.0°C
Quantity 1L
Chemical Name or Material Xylenes
Show More Show Less