Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Xylenes, 98+%, for analysis, mixture of isomers , ACROS Organics™
Click to view available options
Quantity:
1L
5L
Specifications
Assay Percent Range | 98+% |
Linear Formula | C6H4(CH3)2 |
Solubility Information | Solubility in water: 0.1g/L water (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf, methylene chloride, acetone and ether |
Formula Weight | 106.17 |
Percent Purity | 98+% |
Grade | Analysis |
Density | 0.8650g/mL |
Solubility | Solubility in water: 0.1g/L water (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf, methylene chloride, acetone and ether |
Specific Gravity | 0.865 |
Flash Point | 23°C |
Show More |