Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenes, 99%, for biochemistry and histology, mixed isomers with ethylbenzene, AcroS, Acros Organics

Click to view available options
Quantity:
1L
2.5L
Packaging:
AcroSeal™ Glass bottle
1330-20-7
C8H10
106.17
IVSZLXZYQVIEFR-UHFFFAOYSA-N
1,3-xylene
CC1=CC(C)=CC=C1
1330-20-7
C8H10
106.17
IVSZLXZYQVIEFR-UHFFFAOYSA-N
1,3-xylene
CC1=CC(C)=CC=C1
Flash Point 23°C
Viscosity 0.6 mPa.s (20°C)
Assay Percent Range 0.4% max. Toluene (GC)
Linear Formula C6H4(CH3)2
Merck Index 15,10278
Solubility Information Solubility in water: 0.2g/L (20°C). Other solubilities: soluble in methanol,ethanol,2-propanol,dmf,,methylene chloride,acetone and ether
Formula Weight 106.17
Percent Purity 99%
Grade Biochemistry and histology
Density 0.8650g/mL
Packaging AcroSeal™ Glass bottle
Refractive Index 1.4940 to 1.4980
Specific Gravity 0.865
Boiling Point 136.0°C to 140.0°C
Melting Point -34.0°C
Quantity 2.5L
Chemical Name or Material Xylenes
Show More Show Less

GHS H Statement
Harmful if inhaled.
Harmful in contact with skin.
May be fatal if swallowed and enters airways.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
May cause

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Do NOT induce vomiting.

GHS Signal Word: Danger

EINECSNumber : 215-535-7