Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenes, ACS, 98.5%, Alfa Aesar

Manufacturer:  Alfa Aesar 16371AP

Quantity 500mL

 View more versions of this product

Catalog No. AA16371AP


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1330-20-7
>98.5%
106.17
Xylenes
Liquid
(CH3)2C6H4
MFCD00077264
ACS Grade

Specifications

1330-20-7
Liquid
(CH3)2C6H4
MFCD00077264
465°C (869°F)
500mL
ACS Grade
Colorless liquid
>98.5%
106.17
0.86g/cm3
Clear colorless
Xylenes, For ACS analysis

Safety and Handling

SDS & Safety Documents

H226-H312-H332-H315-H303: Flammable liquid and vapour. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. May be harmful if swallowed.

DOTInformation : Hazard Class: 3, Packing Group: III

EINECSNumber : 215-535-7

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit