Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenes, contains ethylbenzene, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
1L
4L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle

Chemical Identifiers

1330-20-7
C6H4(CH3)2

High quality chemicals for laboratory and specialized industrial use

    For Microelectronic Use
Formula Weight 106.17
Packaging Aluminum Bottle
Specific Gravity 1L = 0.86kg
Flash Point 29.00°C
Quantity 4L
Chemical Name or Material Xylenes, contains ethylbenzene
Grade Baker Analyzed™ ACS Reagent