Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenes, extra pure, mixture of isomers, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
10L
Packaging:
Glass bottle
Plastic drum
1330-20-7,100-41-4
C8H10
Dimethylbenzene
1330-20-7,100-41-4
C8H10
Dimethylbenzene
Acidity 0.0003 meq/g max.
Linear Formula C6H4(CH3)2
Formula Weight 106.17
Physical Form Powder
Percent Purity ≥98.0% (mixed isomers) (GC)
Grade Extra Pure
Density 0.8650g/mL
Packaging Plastic drum
Refractive Index 1.4940 to 1.4980
Residue after Evaporation 0.001 % max.
Solubility Solubility in water: <0.1g/L (20°C). Other solubilities: soluble in methanol, ethanol, 2-propanol, dmf,, methylene chloride, acetone and ether
Specific Gravity 0.865
Substances Darkened by H2SO4 Passes test
Water 0.05% Max.
Flash Point 23°C
Viscosity 0.6 mPa.s (20°C)
Boiling Point 136.0°C to 140.0°C
Color Yellow to Beige
Melting Point -34.0°C
Quantity 10L
Chemical Name or Material Xylenes
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Harmful in contact with skin.
Highly flammable liquid and vapor.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning