Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenes (Histological), Fisher Chemical™

Catalog Number p-22629
Click to view available options
Quantity:
1 gal.
4 L
20 L
50 L
200 L
Packaging:
Aluminum Can
BasicPak (Rented)
Poly Bottle
Poly Bottle, F-Style
Steel Drum
Steel Pail
1330-20-7, 100-41-4
C8H10
MFCD00077264
Xylol, Dimethylbenzene
1330-20-7, 100-41-4
C8H10
MFCD00077264
Xylol, Dimethylbenzene
Melting Point -34°C
Boiling Point 136°C
Quantity 50 L
Packaging BasicPak (Rented)
Physical Form Liquid
Linear Formula C6H4(CH3)2
Vapor Pressure 8.29mmHg at 25°C
Formula Weight 106.17
Grade Histological
Chemical Name or Material Xylenes
Hazard Statement WARNING!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Harmful if absorbed through skin or if inhaled. Causes eye, skin, and respiratory tract irritation. Inhalation may cause central nervous system effects. Aspiration hazard if swallowed - can enter
DOTInformation DOT Class 3, : Flammable Liquid
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit