Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenes Laboratory, Macron™

Click to view available options
Quantity:
4L
Packaging:
Glass
Poly
1330-20-7
C6H4(CH3)2
1330-20-7
C6H4(CH3)2
Formula Weight 106.17
Grade Laboratory
Packaging Glass
Quantity 4L
Chemical Name or Material Xylenes