Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenes, Semiconductor Grade, Alfa Aesar™

Manufacturer:  Alfa Aesar 19402K7

Quantity 4L

 View more versions of this product

Catalog No. AA19402K7


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

1330-20-7
(CH3)2C6H4
MFCD00077264
Semiconductor Grade
Liquid
106.17
Xylenes

Specifications

1330-20-7
Liquid
106.17
465°C (869°F)
10 hPa (8mm Hg) at 20°C
4L
Semiconductor Grade
Colorless liquid
(CH3)2C6H4
MFCD00077264
Aromatic
Clear colorless
Xylenes

Safety and Handling

SDS & Safety Documents

H226-H312-H332-H315-H303: Flammable liquid and vapour. Harmful in contact with skin. Harmful if inhaled. Causes skin irritation. May be harmful if swallowed.

DOTInformation : Hazard Class: 3, Packing Group: III

EINECSNumber : 215-535-7

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit