Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenol Orange, BAKER™, J.T.Baker™

Click to view available options
Quantity:
5g
10g
Packaging:
Wide Mouth Amber Glass Bottle
3618-43-7
C31H28N2Na4O13S
3618-43-7
C31H28N2Na4O13S

A grade of chemicals for laboratory use

Formula Weight 760.6
Packaging Wide Mouth Amber Glass Bottle
Quantity 10g
Chemical Name or Material Xylenol Orange