Learn More
Thermo Scientific Chemicals Xylenol Orange tetrasodium salt, ACS
Description
Xylenol Orange tetrasodium salt is used as a photometric reagent and a metal indicator. It is a versatile indicator for EDTA titrations in acidic range. It is a common organic reagent used as an indicator for metal titrations.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | 210°C (decomposition) |
Solubility Information | Soluble in water. |
Formula Weight | 760.59 |
Grade | ACS |
Odor | Odorless |
Quantity | 10 g |
Physical Form | Powder |
Chemical Name or Material | Xylenol Orange tetrasodium salt |
Safety and Handling
EINECSNumber | 222-805-8 |
TSCA | Yes |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.