Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Xylenol Orange Tetrasodium Salt (Certified), Fisher Chemical
Click to view available options
Quantity:
5 g
Packaging:
Glass Bottle
Specifications
Melting Point | 210°C |
Formula Weight | 760.59 |
Grade | Certified |
Packaging | Glass Bottle |
Suitability | Pass Test |
Color | Red |
Quantity | 5 g |
Physical Form | Solid |
Chemical Name or Material | Xylenol Orange Tetrasodium Salt |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction