Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Xylenol Orange Triturate R, Ricca Chemical

Click to view available options
Quantity:
100g
7757-79-1,3618-43-7
KNO3
101.102
FGIUAXJPYTZDNR-UHFFFAOYSA-N
24434
CHEBI:63043
potassium;nitrate
[N+](=O)([O-])[O-].[K+]
7757-79-1,3618-43-7
KNO3
101.102
FGIUAXJPYTZDNR-UHFFFAOYSA-N
24434
CHEBI:63043
potassium;nitrate
[N+](=O)([O-])[O-].[K+]
CAS Min % 97.0,0.98
CAS Max % 100.0,1.02
Physical Form Powder
Concentration or Composition Notes 1% (w/w)
Solvent or Matrix Potassium Nitrate
Color Red to Brown
Quantity 100g
Chemical Name or Material Xylenol Orange Triturate, Reagent